Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTF-1/ NKX2-1 Antibody (6C11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | TTF-1 / NKX2-1 |
---|---|
Clone | 6C11 |
Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
TTF-1 / NKX2-1 | |
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
A synthesized peptide derived from Human TTF-1/NKX2-1 [UniProt P43699] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
6C11 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Membrane Trafficking and Chaperones, Tumor Biomarkers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
7080 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title