Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTF-1/ NKX2-1 Antibody (8G7G3/1 + NX2.1/690), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247773AF405
Description
TTF-1 / NKX2-1 Monoclonal specifically detects TTF-1 / NKX2-1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
TTF-1 / NKX2-1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NKX2-1 | |
Rat full length TTF-1/ NKX2-1 recombinant protein (8G7G3/1); Recombinant TTF-1/ NKX2-1 protein (NX2.1/690) (Uniprot: P43699) | |
0.1 mL | |
Primary | |
Recognizes a protein of 40kDa, identified as Thyroid transcription factor-1 (TTF-1). TTF-1 is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. Anti-TTF-1 is useful in differentiating primary adenocarcinoma of the lung from metastatic carcinomas originating in the breast, mediastinal germ cell tumors, and malignant mesothelioma. It can also be used to differentiate small cell lung carcinoma from lymphoid infiltrates.Loss of TTF-1 expression in non-small cell lung carcinoma has been associated with aggressive behavior of such neoplasms. TTF-1 reactivity is also seen in thyroid malignancies. | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
8G7G3/1 + NX2.1/690 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence | |
benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
Mouse | |
Protein A or G purified | |
Membrane Trafficking and Chaperones, Tumor Biomarkers | |
7080 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction