Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTF-1/ NKX2-1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | TTF-1 / NKX2-1 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TTF-1 / NKX2-1 Polyclonal specifically detects TTF-1 / NKX2-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TTF-1 / NKX2-1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
NKX2-1 | |
IgG | |
Affinity Purified | |
41, 45 kDa |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Membrane Trafficking and Chaperones, Tumor Biomarkers | |
PBS with 0.02% Sodium Azide | |
7080 | |
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human NKX2-1. The immunogen is located within the first 50 amino acids of NKX2-1. Amino Acid Squence: LSPLEESYKKVGMEGG | |
Primary | |
NKX2-1 antibody is predicted to not cross-react with other NK2 homeobox family members. At least three isoforms of NKX2-1 are known to exist; this antibody will detect all three. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title