Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTF-1/ NKX2-1 Antibody (TTF1/8143R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320240100UG
Description
TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)Specifications
TTF-1 / NKX2-1 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
Recombinant fragment (around aa1-400) of human TTF-1/ NKX2-1 protein (exact sequence is proprietary) | |
100 μg | |
Membrane Trafficking and Chaperones, Tumor Biomarkers | |
7080 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
TTF1/8143R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction