Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTF1 Mouse anti-Human, Porcine, Clone: 2C8F3, Proteintech
SDP

Catalog No. 501736474 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6474 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6474 Supplier Proteintech Group Inc Supplier No. 660341IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen TTF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2C8F3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NKX2-1
Gene Accession No. P43699
Gene Alias BHC, Homeobox protein Nkx 2.1, NK 2, NK2 homeobox 1, NKX2 1, NKX2.1, NKX2-1, NKX2A, TEBP, Thyroid nuclear factor 1, Thyroid transcription factor 1, TITF1, TTF 1, TTF1
Gene Symbols NKX2-1
Host Species Mouse
Immunogen Peptide
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100154384, 7080
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.