Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTF1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501562408
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2408 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2408 Supplier Bethyl Laboratories, Inc Supplier No. A302361AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Thyroid transcription factor-1 (TTF-1), also known as NK2 homeobox 1(NKX2-1), is a member of the NKx2 family of homeodomain transcription factors. It binds and activates the promoter of thyroid specific genes such as thyroglobulin, thyroperoxidase, and thyrotropin receptor. TTF-1 is important in the maintece of thyroid differentiation and lung development. It is expressed in epithelial cells of the thyroid gland and the lung, and tumors derived from such epithelial cells. TTF-1 is usually found in primary lung adenocarcinomas, small cell carcinomas and thyroid cancer. It is absent in mesotheliomas, breast cancer and colon cancer.

Specifications

Antigen TTF1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene TTF1
Gene Accession No. Q15361
Gene Alias TTF-1, TTF-I, transcription termination factor 1
Gene Symbols TTF1
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7270
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.