Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTF1 (thyroid transcription factor 1) Monoclonal Antibody (SP141), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 604870

Description
Positive control: Thyroid Cellular location: Nucleus.
TTF1 (thyroid transcription factor 1) Monoclonal antibody specifically detects TTF1 (thyroid transcription factor 1) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
TTF1 (thyroid transcription factor 1) | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
Q15361 | |
Mouse | |
Protein A | |
RUO | |
7270 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
SP141 | |
Unconjugated | |
Ttf1 | |
AV245725; BCH; BHC; Homeobox protein NK-2 homolog A; homeobox protein Nkx-2.1; NK-2; NK2 homeobox 1; NK-2 homolog A; NKX2.1; NKX2-1; NKX2A; NMTC1; RGD1565673; RNA polymerase I termination factor; RNA polymerase I transcription termination factor; T/EBP; TEBP; Thyroid nuclear factor 1; thyroid transcription factor 1; thyroid transcription factor 1-like; thyroid-specific enhancer-binding protein; TITF1; Transcription termination factor 1; transcription termination factor I; transcription termination factor, RNA polymerase I; TTF1; TTF-1; TTF-I | |
A recombinant protein of human TTF1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction