Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596789
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596789 Supplier Invitrogen™ Supplier No. PA596789
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEEVRCPEHG TFCFLKTGVR DGPNKGKSFY VCRADTCSFV RATDIPVSHC LLHEDFVVEL QGLLLPQDKK EYRLFFRCIR SKAEGKRWCG SIPWQDPDSK EHSVSNKSQH ASETFHHSSN WLRNPFKVLD KNQEPALWKQ LIKGEGEEKK ADKKQREKGD QLFDQKKEQK PEMMEKDLSS GLVPKKKQSV VQEKKQEEGA EIQCEAETGG THKRDFSEIK SQQCQGNELT RPSASSQEKS SGKSQDVQRE SEPLREKVTQ LLPQNVHSHN; Positive Samples: MCF7, HL-60, SKOV3, Jurkat; Cellular Location: Cytoplasm, Nucleus.

This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTF2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TTF2
Gene Accession No. Q9UNY4
Gene Alias 4632434F22Rik; 8030447N19; AV218430; F2; HuF2; human factor 2; lodestar homolog; lodestar protein; RNA polymerase II termination factor; Transcription release factor 2; transcription termination factor 2; transcription termination factor, RNA polymerase II; Ttf2; ZGRF6; zinc finger, GRF-type containing 6
Gene Symbols TTF2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human TTF2 (NP_0035853).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8458
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.