Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTI1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566987
Description
Immunogen sequence: ERLDLGEGDLN KVADACLIYL SVKQPVKLQE AARSVFLHLM KVDPDSTWFL LNELYCPVQF TPPHPSLHPV QLHGASGQQN PYTT Highest antigen sequence identity to the following orthologs - mouse 85%, rat 83%.
Regulator of the DNA damage response (DDR). Part of the TTT complex that is required to stabilize protein levels of the phosphatidylinositol 3-kinase-related protein kinase (PIKK) family proteins. The TTT complex is involved in the cellular resistance to DNA damage stresses, like ionizing radiation (IR), ultraviolet (UV) and mitomycin C (MMC). Together with the TTT complex and HSP90 may participate in the proper folding of newly synthesized PIKKs. Promotes assembly, stabilizes and maintains the activity of mTORC1 and mTORC2 complexes, which regulate cell growth and survival in response to nutrient and hormonal signals.
Specifications
TTI1 | |
Polyclonal | |
Unconjugated | |
TTI1 | |
2610036D13Rik; AI449463; KIAA0406; Protein SMG10; SMG10; smg-10; smg-10 homolog, nonsense mediated mRNA decay factor; Tel two interacting protein 1; Tel2 interacting protein 1 homolog; TEL2-interacting protein 1 homolog; TELO2 interacting protein 1; TELO2-interacting protein 1 homolog; TTI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9675 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43156 | |
TTI1 | |
Recombinant Human TTI1. Recombinant protein control fragment (Product #RP-107631). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction