Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554812
Description
Immunogen sequence: LLGKVETAKN SLVGPAWQTG LHHLAGPVYI FAITHSLEQP WTTPRSREVA REVLTSLLQV TECGSVAGFL HGENEDEKG Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 70%.
The DNA replication (origin) licensing system is necessary for duplicating the genome in each cell cycle. Geminin is a protein that prevents DNA replication by blocking the MCM complex incorporation into prereplication complex (pre-RC). It is absent during G1 phase and accumulates during S, G2, and M phases of mitosis. It is then degraded during the metaphase-anaphase trasition allowing replication to occur during subsequent cycles. Geminin has also been implicated in maintaining integrity of the male germline genome.
Specifications
TTI2 | |
Polyclonal | |
Unconjugated | |
TTI2 | |
C8orf41; FLJ23263; MRT39; RGD1310414; Tel2 interacting protein 2 homolog; TELO2 interacting protein 2; TELO2-interacting protein 2; TTI2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80185 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NXR4 | |
TTI2 | |
Recombinant protein corresponding to Human TTI2. Recombinant protein control fragment (Product #RP-93782). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction