Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556269
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556269 Supplier Invitrogen™ Supplier No. PA556269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEDSNLSEEL SHSAFGQAFS KILHCLARPE ARRGNVKDAV LKDLGDLIEA TGFDRLFEGT GARLRGMPET LGQVAKALEK Y Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 57%.

The DNA replication (origin) licensing system is necessary for duplicating the genome in each cell cycle. Geminin is a protein that prevents DNA replication by blocking the MCM complex incorporation into prereplication complex (pre-RC). It is absent during G1 phase and accumulates during S, G2, and M phases of mitosis. It is then degraded during the metaphase-anaphase trasition allowing replication to occur during subsequent cycles. Geminin has also been implicated in maintaining integrity of the male germline genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTI2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTI2
Gene Accession No. Q6NXR4
Gene Alias C8orf41; FLJ23263; MRT39; RGD1310414; Tel2 interacting protein 2 homolog; TELO2 interacting protein 2; TELO2-interacting protein 2; TTI2
Gene Symbols TTI2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTI2. Recombinant protein control fragment (Product #RP-93783).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80185
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.