Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598544
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598544 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598544 Supplier Invitrogen™ Supplier No. PA598544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTI2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The DNA replication (origin) licensing system is necessary for duplicating the genome in each cell cycle. Geminin is a protein that prevents DNA replication by blocking the MCM complex incorporation into prereplication complex (pre-RC). It is absent during G1 phase and accumulates during S, G2, and M phases of mitosis. It is then degraded during the metaphase-anaphase trasition allowing replication to occur during subsequent cycles. Geminin has also been implicated in maintaining integrity of the male germline genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTI2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TTI2
Gene Accession No. Q66H56, Q6NXR4
Gene Alias C8orf41; FLJ23263; MRT39; RGD1310414; Tel2 interacting protein 2 homolog; TELO2 interacting protein 2; TELO2-interacting protein 2; TTI2
Gene Symbols TTI2
Host Species Rabbit
Immunogen Recombinant Human TELO2-interacting protein 2 protein (159-264AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290811, 80185
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.