Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTK Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298832AF532
Description
TTK Polyclonal specifically detects TTK in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TTK | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
CT96, dual specificity protein kinase TTK, EC 2.7.12.1, ESK, FLJ38280, monopolar spindle 1-like 1, MPS1, MPS1L1PYTcancer/testis antigen 96, phosphotyrosine picked threonine kinase, Phosphotyrosine picked threonine-protein kinase, TTK protein kinase | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human TTK. | |
0.1 mL | |
Breast Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Protein Kinase, Protein Phosphatase, Stem Cell Markers | |
7272 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction