Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116969 Supplier Invitrogen™ Supplier No. PA5116969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa Immunogen sequence: SSKVFQVLNE KKQIYAIKYV NLEEADNQTL DSYRNEIAYL NKLQQHSDKI IRLYDYEITD QYIYMVMECG NIDLNSWLKK KKSIDPWERK SYWKNMLEAV HTIHQHGIVH SDLKPANFLI VDGMLKLIDF GIANQMQPDT TSVVKDSQVG TVNYMPPEAI KDMSSSRENG KSKSKISPKS DVWSLGCILY YMTYGKTPFQ QIINQISKLH AIIDPNHEIE FPDIPEKDLQ DVLKCCLKRD PKQRISIPEL LAHP.

TTK is a serine/threonine kinase closely related to SPK1, PIM 1 and CDC2 serine/threonine kinases. TTK is a dual-specificity kinase, which can also phosphorylate tyrosine. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. TTK encodes an alpha isoform of the constant regulatory subunit A. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTK
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TTK
Gene Accession No. P33981, P35761
Gene Alias AL022661; cancer/testis antigen 96; CT96; Dual specificity protein kinase TTK; esk; Esk1; monopolar spindle 1 kinase; monopolar spindle 1-like 1; MPH1; MPS1; MPS1L1; OTTHUMP00000016778; OTTHUMP00000220743; phosphotyrosine picked threonine kinase; phosphotyrosine picked threonine-protein kinase; PYT; TTK; Ttk protein kinase
Gene Symbols TTK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 536-789 of human TTK (NP_0033092).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22137, 315852, 7272
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.