Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119482
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119482 200 μL
PIPA5121518 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119482 Supplier Invitrogen™ Supplier No. PA5119482
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TTK Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

TTK is a serine/threonine kinase closely related to SPK1, PIM 1 and CDC2 serine/threonine kinases. TTK is a dual-specificity kinase, which can also phosphorylate tyrosine. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. TTK encodes an alpha isoform of the constant regulatory subunit A. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TTK
Gene Accession No. P33981, P35761
Gene Alias AL022661; cancer/testis antigen 96; CT96; Dual specificity protein kinase TTK; esk; Esk1; monopolar spindle 1 kinase; monopolar spindle 1-like 1; MPH1; MPS1; MPS1L1; OTTHUMP00000016778; OTTHUMP00000220743; phosphotyrosine picked threonine kinase; phosphotyrosine picked threonine-protein kinase; PYT; TTK; Ttk protein kinase
Gene Symbols TTK
Host Species Rabbit
Immunogen Recombinant protein TTK Protein Kinase. The antigen corresponds to amino acid range 532-786 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22137, 7272
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.