Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTL Antibody (OTI3G5), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274668C
Description
TTL Monoclonal specifically detects TTL in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TTL | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TTL | |
Full length recombinant protein of human TTL (NP_714923) produced in HEK293T cells. | |
0.1 mL | |
150465.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3G5 | |
Western Blot | |
EC 6.3.2.25,2410003M22Rik, MGC46235, tubulin tyrosine ligase, tubulin-tyrosine ligase, tubulin--tyrosine ligase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction