Learn More
Invitrogen™ TTL Monoclonal Antibody (1B2)

Description
Peptide Sequence: LESSILLQIK HIIRNCLLSV EPAISTKHLP YQSFQLFGFD FMVDEELKVW LIEVNGAPAC AQKLYAELCQ GIVDIAISSV FPPPDVEQPQ TQPAAFIKL.
Specifications
Specifications
| Antigen | TTL |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1B2 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TTL |
| Gene Accession No. | Q8NG68 |
| Gene Alias | 2410003M22Rik; 2700049H19Rik; AI848570; Ttl; tubulin tyrosine ligase; tubulin-tyrosine ligase; tubulin--tyrosine ligase |
| Gene Symbols | TTL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.