Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTL Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E5F8, Proteintech
SDP

Catalog No. 501736513 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6513 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6513 Supplier Proteintech Group Inc Supplier No. 660761IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.

Specifications

Antigen TTL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2E5F8
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TTL
Gene Accession No. P38160, P38585, Q8NG68, Q9QXJ0
Gene Alias TTL, Tubulin tyrosine ligase, tubulin tyrosine ligase
Gene Symbols Ttl
Host Species Mouse
Immunogen TTL Fusion Protein Ag4708
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150465, 171572, 445530, 69737
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.