Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566871
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566871 Supplier Invitrogen™ Supplier No. PA566871
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQIKHIIRNCL LSVEPAISTK HLPYQSFQLF GFDFMVDEEL KVWLIEVNGA PACAQKLYAE LCQGIVDIAI SSVFPPPDVE QPQTQP Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTL
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTL
Gene Accession No. Q8NG68
Gene Alias 2410003M22Rik; 2700049H19Rik; AI848570; Ttl; tubulin tyrosine ligase; tubulin-tyrosine ligase; tubulin--tyrosine ligase
Gene Symbols TTL
Host Species Rabbit
Immunogen Recombinant Human TTL. Recombinant protein control fragment (Product #RP-107522).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150465
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.