Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561731
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561731 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561731 Supplier Invitrogen™ Supplier No. PA561731
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLTSSTANDR ILKYNLINDT LNIAVPNGEI PDCKWNKSPP KEVLGNYEIL YDEELAQGDG ADRELRSRQG QSLGPRAGRS RDSGRAVLTT WK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

Catalytic subunit of the neuronal tubulin polyglutamylase complex. Modifies alpha- and beta-tubulin, generating side chains of glutamate on the gamma-carboxyl groups of specific glutamate residues within the C-terminal tail of alpha- and beta-tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTLL1
Gene Accession No. O95922
Gene Alias 6330444E16Rik; AV014541; bM121M7.1 (novel protein (ortholog of human tubulin tyrosine ligase-like 1 (TTLL1)), isoform 1); C22orf7; catalytic subunit of neural tubulin polyglutamylase; HS323M22B; p49; PGs3; Probable tubulin polyglutamylase TTLL1; TTLL1; Tubulin polyglutamylase complex subunit 3; tubulin tyrosine ligase like 1; tubulin tyrosine ligase-like 1; tubulin tyrosine ligase-like family member 1; tubulin tyrosine ligase-like family, member 1; tubulin-tyrosine ligase; tubulin--tyrosine ligase-like protein 1
Gene Symbols TTLL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTLL1. Recombinant protein control fragment (Product #RP-100904).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.