Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546070 Supplier Invitrogen™ Supplier No. PA546070
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QVLQRPPPTL PPSKPKPVQG LCPHGKPRDK GRSCKRSSGH GSGENGSQRP Sequence homology: Human: 100%; Rabbit: 92%.

TTLL11 is a polyglutamase which preferentially modifies alpha-tubulin. It is involved in the side-chain elongation step of the polyglutamylation reaction rather than in the initiation step. TTLL11 is required for CCSAP localization to both spindle and cilia microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TTLL11
Gene Accession No. Q8NHH1
Gene Alias bA244O19.1; C9orf20; TTLL11; Tubulin polyglutamylase TTLL11; tubulin tyrosine ligase like 11; tubulin tyrosine ligase like11; tubulin tyrosine ligase-like family member 11; tubulin tyrosine ligase-like family, member 11; tubulin--tyrosine ligase-like protein 11
Gene Symbols TTLL11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TTLL11 (aa 169-218).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158135
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.