Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTLL12 Antibody (OTI7F11), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274676F
Description
TTLL12 Monoclonal specifically detects TTLL12 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TTLL12 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TTLL12 | |
Full length recombinant protein of human TTLL12 (NP_055955) produced in HEK293T cells. | |
0.1 mL | |
23170.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI7F11 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
dJ526I14.2, KIAA0153FLJ41795, tubulin tyrosine ligase-like family, member 12, tubulin--tyrosine ligase-like protein 12 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction