Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117122
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117122 Supplier Invitrogen™ Supplier No. PA5117122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Jurkat, HL-60, Mouse brain, Mouse spinal cord, Mouse heart, Rat brain Immunogen sequence: MEAERGPERR PAERSSPGQT PEEGAQALAE FAALHGPALR ASGVPERYWG RLLHKLEHEV FDAGEVFGIM QVEEVEEEED EAAREVRKQQ PNPGNELCYK VIVTRESGLQ AAHPNSIFLI DHAWTCRVEH ARQQLQQVPG LLHRMANLMG IEFHGELPST EAVALVLEEM WKFNQTYQLA HGTAEEKMPV WYIMDEFGSR IQHADVPSFA TAPFFYMPQQ VAYTLLWPLR DLDTGEEVTR DFAYGETDPL IRKCMLLPWA PTDMLDLSSC TPEPPAEHYQ AILEENKEKL PLDINPVVHP HGHIFKVYTD VQQVASSLTH PRFTLTQSEA DADILFNFSH FKDYRKLSQE RPGVLLNQFP CENLLTVKDC LASIARRAGG PEGPPWLPRT FNLRTELPQF VSYFQQRERW GEDNHWICKP WNLARSLDTH VTKSLHSIIR HRESTPKVVS.

TTLL12 (tubulin tyrosine ligase-like family, member 12) is a 644 amino acid protein that contains one TTL domain. Of the fourteen members of the TTLL family that modify tubulin, TTLL12 is the least characterized member. TTLL12 is highly expressed in a multitude of metastatic prostate cancer cell lines, therefore, it is considered a target for tumor therapy. Overexpression of TTLL12 is suggested to alter chromosome ploidy, whereas downregulation of TTLL12 influence several post-translational modifications of tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL12
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TTLL12
Gene Accession No. Q14166, Q3UDE2
Gene Alias BC055368; D430005B17; dJ526I14.2; Inactive tubulin--tyrosine ligase-like protein 12; KIAA0153; RGD1305319; TTL domain protein; ttll12; tubulin tyrosine ligase like 12; tubulin tyrosine ligase-like family member 12; tubulin tyrosine ligase-like family, member 12; tubulin--tyrosine ligase-like protein 12
Gene Symbols TTLL12
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-450 of human TTLL12 (NP_0559551).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223723, 23170, 300105
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.