Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144176
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144176 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144176 Supplier Invitrogen™ Supplier No. PA5144176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTLL2 Polyclonal Antibody for IHC (P), ELISA

Probable tubulin polyglutamylase that generates side chains of glutamate on the gamma-carboxyl group of specific glutamate residues within the C-terminal tail of target proteins (By similarity). Similar to TTLL1, may acquire enzymatic activity only in complex with other proteins as it is most likely lacking domains important for autonomous activity (By similarity). Probably involved in the side-chain initiation step of the polyglutamylation reaction rather than the elongation step (By similarity).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene TTLL2
Gene Accession No. Q9BWV7
Gene Alias C6orf104
Gene Symbols TTLL2
Host Species Rabbit
Immunogen Recombinant Human Probable tubulin polyglutamylase TTLL2 protein (401-592AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83887
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.