Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTLL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570598
Description
This target displays homology in the following species: Human: 100%.
TTLL3 gene ontology annotations related to this gene include protein polyglycylation.
Specifications
TTLL3 | |
Polyclonal | |
Unconjugated | |
TTLL3 | |
HOTTL; PRO0207; TTLL3; Tubulin monoglycylase TTLL3; tubulin tyrosine ligase like 3; tubulin tyrosine ligase-like family member 3; tubulin tyrosine ligase-like family, member 3; tubulin--tyrosine ligase-like protein 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26140 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y4R7 | |
TTLL3 | |
synthetic peptide directed towards the C-terminal region of TTLL3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction