Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555500 Supplier Invitrogen™ Supplier No. PA555500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGLEDCCSRD ENEEEEGDSE CSSLSAVSPS ESVAMISRSC MEILTKPLSN HEKVVRPALI YSLFPNVPPT IYFGTRDERV EKLPWEQRKL LRWKMSTVTP NIVKQTIGRS HFKISKRNDD WLGCWGHHMK SPSFRSI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Glutamylase which preferentially modifies beta-tubulin and non-tubulin proteins, such as NAP1L1, NAP1L4 and CGAS. Involved in the side-chain initiation step of the polyglutamylation reaction rather than in the elongation step. Involved in formation of short side-chains. Mediates initiation of polyglutamylation of nucleosome assembly proteins NAP1L1 and NAP1L4. Also acts as a monoglutamylase: generates monoglutamylation of CGAS, leading to impair the nucleotidyltransferase activity of CGAS. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTLL4
Gene Accession No. Q14679
Gene Alias KIAA0173; TTLL4; Tubulin polyglutamylase TTLL4; tubulin tyrosine ligase like 4; tubulin tyrosine ligase-like family member 4; tubulin tyrosine ligase-like family, member 4; tubulin--tyrosine ligase-like protein 4
Gene Symbols TTLL4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTLL4. Recombinant protein control fragment (Product #RP-91633).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9654
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.