Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142543
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142543 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142543 Supplier Invitrogen™ Supplier No. PA5142543
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

Glutamylase which preferentially modifies beta-tubulin and non-tubulin proteins, such as NAP1L1, NAP1L4 and CGAS. Involved in the side-chain initiation step of the polyglutamylation reaction rather than in the elongation step. Involved in formation of short side-chains. Mediates initiation of polyglutamylation of nucleosome assembly proteins NAP1L1 and NAP1L4. Also acts as a monoglutamylase: generates monoglutamylation of CGAS, leading to impair the nucleotidyltransferase activity of CGAS. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene TTLL4
Gene Accession No. Q14679
Gene Alias KIAA0173; TTLL4; Tubulin polyglutamylase TTLL4; tubulin tyrosine ligase like 4; tubulin tyrosine ligase-like family member 4; tubulin tyrosine ligase-like family, member 4; tubulin--tyrosine ligase-like protein 4
Gene Symbols TTLL4
Host Species Goat
Immunogen Peptide with sequence C-DSQDPTKEIRFTE.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9654
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.