Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556938
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556938 Supplier Invitrogen™ Supplier No. PA556938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NKHHSGIAKT QKEGEDASLY SKRYNQSMVT AELQRLAEKQ AARQYSPSSH INLLTQQVTN LNLATGIINR SSASAPPTLR PIISPSGP Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.

Polyglutamylase which preferentially modifies alpha-tubulin. Involved in the side-chain initiation step of the polyglutamylation reaction rather than in the elongation step. Required for CCSAP localization to both spindle and cilia microtubules. Increases the effects of NCOA2 in glucocorticoid receptor-mediated repression and induction and in androgen receptor-mediated induction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTLL5
Gene Accession No. Q6EMB2
Gene Alias CORD19; KIAA0998; SRC1 and TIF2 associated binding protein; SRC1 and TIF2-associated modulatory protein; STAMP; TTLL5; Tubulin polyglutamylase TTLL5; tubulin tyrosine ligase like 5; tubulin tyrosine ligase-like family member 5; tubulin tyrosine ligase-like family, member 5; Tubulin--tyrosine ligase-like protein 5
Gene Symbols TTLL5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTLL5. Recombinant protein control fragment (Product #RP-95623).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23093
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.