Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571379 Supplier Invitrogen™ Supplier No. PA571379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTLL5 Polyclonal Antibody for Western Blot

Polyglutamylase which preferentially modifies alpha-tubulin. Involved in the side-chain initiation step of the polyglutamylation reaction rather than in the elongation step. Required for CCSAP localization to both spindle and cilia microtubules. Increases the effects of NCOA2 in glucocorticoid receptor-mediated repression and induction and in androgen receptor-mediated induction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TTLL5
Gene Accession No. Q6EMB2
Gene Alias CORD19; KIAA0998; SRC1 and TIF2 associated binding protein; SRC1 and TIF2-associated modulatory protein; STAMP; TTLL5; Tubulin polyglutamylase TTLL5; tubulin tyrosine ligase like 5; tubulin tyrosine ligase-like family member 5; tubulin tyrosine ligase-like family, member 5; Tubulin--tyrosine ligase-like protein 5
Gene Symbols TTLL5
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human TTLL5.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23093
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.