Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562599 Supplier Invitrogen™ Supplier No. PA562599
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLKSDMNKPH LISELLTKLQ LSGKLSFFPA HYNPKLGMNN LSQNPSLPGE CHSRSDSSGE KRQLDVSSLL LQSPQSYNVT LRDLLVIATP AQLDPRPCRS HASAMRDPCM QDQE Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 41%.

Predicted to enable tubulin binding activity and tubulin-glutamic acid ligase activity. Predicted to be involved in microtubule cytoskeleton organization and protein polyglutamylation. Predicted to act upstream of or within microtubule bundle formation; microtubule severing; and positive regulation of cilium movement. Located in ciliary basal body.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTLL6
Gene Accession No. Q8N841
Gene Alias 4932418K24Rik; D11Moh43e; D11Moh44e; polyglutamylase; RGD1311922; t8130b59; TTL.6; TTLL6; Ttll6-ps1; Tubulin polyglutamylase TTLL6; tubulin tyrosine ligase like 6; tubulin tyrosine ligase-like family member 6; tubulin tyrosine ligase-like family, member 6; tubulin tyrosine ligase-like family, member 6, pseudogene 1; tubulin--tyrosine ligase-like protein 6
Gene Symbols TTLL6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTLL6. Recombinant protein control fragment (Product #RP-104130).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.