Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598617
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598617 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598617 Supplier Invitrogen™ Supplier No. PA598617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TTLL6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Predicted to enable tubulin binding activity and tubulin-glutamic acid ligase activity. Predicted to be involved in microtubule cytoskeleton organization and protein polyglutamylation. Predicted to act upstream of or within microtubule bundle formation; microtubule severing; and positive regulation of cilium movement. Located in ciliary basal body.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TTLL6
Gene Accession No. Q8N841
Gene Alias 4932418K24Rik; D11Moh43e; D11Moh44e; polyglutamylase; RGD1311922; t8130b59; TTL.6; TTLL6; Ttll6-ps1; Tubulin polyglutamylase TTLL6; tubulin tyrosine ligase like 6; tubulin tyrosine ligase-like family member 6; tubulin tyrosine ligase-like family, member 6; tubulin tyrosine ligase-like family, member 6, pseudogene 1; tubulin--tyrosine ligase-like protein 6
Gene Symbols TTLL6
Host Species Rabbit
Immunogen Recombinant Human Tubulin polyglutamylase TTLL6 protein (121-310AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284076
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.