Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566247 Supplier Invitrogen™ Supplier No. PA566247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WNCFCDSGSSW ESIFNKSPEV VTPLQLQCCQ RLVELCKQCL LVVYKYATDK RGSLSGIGPD WGNSRYLLPG STQFFLRTPT YNLKYNSPGM TRSNV Highest antigen sequence identity to the following orthologs - mouse 88%, rat 86%.

Polyglutamylase which preferentially modifies beta-tubulin (PubMed:25959773). Mediates both ATP-dependent initiation and elongation of polyglutamylation of microtubules (PubMed:25959773). Required for neurite growth; responsible for the strong increase in tubulin polyglutamylation during postnatal neuronal maturation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTLL7
Gene Accession No. Q6ZT98
Gene Alias 1110049N09Rik; 4921517B04Rik; C630030B20; mTTLL7; RP5-836J3.2; Testis development protein NYD-SP30; TTLL7; Tubulin polyglutamylase TTLL7; tubulin tyrosine ligase like 7; tubulin tyrosine ligase-like family member 7; tubulin tyrosine ligase-like family, member 7; tubulin--tyrosine ligase-like protein 7
Gene Symbols TTLL7
Host Species Rabbit
Immunogen Recombinant Human TTLL7. Recombinant protein control fragment (Product #RP-106926).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79739
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.