Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTLL7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598547
Description
TTLL7 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Polyglutamylase which preferentially modifies beta-tubulin (PubMed:25959773). Mediates both ATP-dependent initiation and elongation of polyglutamylation of microtubules (PubMed:25959773). Required for neurite growth; responsible for the strong increase in tubulin polyglutamylation during postnatal neuronal maturation. [UniProt].
Specifications
TTLL7 | |
Polyclonal | |
Unconjugated | |
TTLL7 | |
1110049N09Rik; 4921517B04Rik; C630030B20; mTTLL7; RP5-836J3.2; Testis development protein NYD-SP30; TTLL7; Tubulin polyglutamylase TTLL7; tubulin tyrosine ligase like 7; tubulin tyrosine ligase-like family member 7; tubulin tyrosine ligase-like family, member 7; tubulin--tyrosine ligase-like protein 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79739 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6ZT98 | |
TTLL7 | |
Recombinant Human Tubulin polyglutamylase TTLL7 protein (1-260AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction