Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531807 Supplier Invitrogen™ Supplier No. PA531807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Predicted reactivity: Human (99%), Mouse (94%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Polyglutamylase which preferentially modifies beta-tubulin (PubMed:25959773). Mediates both ATP-dependent initiation and elongation of polyglutamylation of microtubules (PubMed:25959773). Required for neurite growth; responsible for the strong increase in tubulin polyglutamylation during postnatal neuronal maturation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL7
Applications Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene TTLL7
Gene Accession No. A4Q9F0, Q6ZT98
Gene Alias 1110049N09Rik; 4921517B04Rik; C630030B20; mTTLL7; RP5-836J3.2; Testis development protein NYD-SP30; TTLL7; Tubulin polyglutamylase TTLL7; tubulin tyrosine ligase like 7; tubulin tyrosine ligase-like family member 7; tubulin tyrosine ligase-like family, member 7; tubulin--tyrosine ligase-like protein 7
Gene Symbols TTLL7
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TTLL7. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70892, 79739
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.