Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTLL8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553512 Supplier Invitrogen™ Supplier No. PA553512
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLEIGLCVNM RSLPWYVPAN PDSFFPRCYS LCTESEQQEF LEDFRRTMAS SILKWVVSHQ SCSRSSRSKP RDQREEAGSS DLSSRQDAEN AEAKLRGLPG QLVDIACKVC QAYLGQLEHE DIDTSADAVE DLTEAEWEDL TQQYYSLVHG DAFISNSRNY FSQCQALLNR ITSVNPQTDI DGLRNIWII Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 59%.

Monoglycylase which modifies both tubulin and non-tubulin proteins, generating side chains of glycine on the gamma-carboxyl groups of specific glutamate residues of target proteins. Monoglycylates tubulin, with a preference for alpha-tubulin toward beta-tubulin. Has the ability to modify non-tubulin proteins such as ANP32A, ANP32B, SET and NCL. Involved in the side-chain initiation step of the glycylation reaction by adding a single glycine chain to generate monoglycine side chains. Not involved in elongation step of the polyglycylation reaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTLL8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTLL8
Gene Accession No. A6PVC2
Gene Alias protein monoglycylase TTLL8; TTLL8; tubulin tyrosine ligase like 8; tubulin tyrosine ligase-like family member 8; tubulin tyrosine ligase-like family, member 8; tubulin--tyrosine ligase-like protein 8
Gene Symbols TTLL8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTLL8. Recombinant protein control fragment (Product #RP-91398).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164714
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.