Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTLL9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TTLL9 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TTLL9 Polyclonal specifically detects TTLL9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TTLL9 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
164395 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GITWIMKPVARSQGKGIFLFRRLKDIVDWRKDTRSSDDQKDDIPVENYVAQRYIE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
C20orf125, chromosome 20 open reading frame 125, dJ310O13.1, EC 6.-, MGC120486, MGC120487, probable tubulin polyglutamylase TTLL9, tubulin tyrosine ligase-like family, member 9, Tubulin--tyrosine ligase-like protein 9 | |
TTLL9 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title