Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TTP Antibody (OTI8B5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274681H
Description
TTP Monoclonal specifically detects TTP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TTP | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ZFP36 | |
Full length recombinant protein of human TTP (NP_003398) produced in HEK293T cells. | |
0.1 mL | |
7538.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI8B5 | |
Western Blot, Immunohistochemistry | |
EZNFTranscription factor 17, HKL1Zinc finger protein eZNF, KID-1, KID1TCF-17, TCF17, zinc finger protein 354A | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction