Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TTR mutant (Y78F) Mouse anti-Human, Clone: AD7, MilliporeSigma™

Catalog No. MABN230725 Shop All MilliporeSigma Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN230725 25 μg
MABN2307100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. MABN230725 missing translation for 'mfr' MilliporeSigma missing translation for 'supplierNo' MABN230725UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TTR mutant (Y78F) Monoclonal antibody specifically detects TTR mutant (Y78F) in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTR mutant (Y78F)
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AD7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Recombinant human Transthyretin with Y78F mutation.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.