Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTYH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534394
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534394 Supplier Invitrogen™ Supplier No. PA534394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is Raji cell lysate. PA5-34394 can be used with blocking peptide PEP-1436.

TTYH1 is a member of the tweety family of proteins, a family of chloride anion channels containing five transmembrane regions. TTYH1 is a Ca2+-independent, volume-sensitive large conductance chloride (Cl-) channel. TTYH1 is primarily expressed in neural tissue and upregulated in astrocytoma, glioma, and several other cancers. Recent experiments have shown that TTYH1 is an integral endoplasmic reticulum (ER) membrane protein involved in cell proliferation and is thought to play an essential role in embryonic cell growth, possibly through the Ca2+ storage/release process in ER membranes during early development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTYH1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TTYH1
Gene Accession No. Q9H313
Gene Alias 4930459B04Rik; 6330408P11Rik; hTTY1; mTTY1; Protein tweety homolog 1; tty; TTY1; TTYH1; tweety family member 1; tweety homolog 1; tweety homolog 1 (Drosophila)
Gene Symbols TTYH1
Host Species Rabbit
Immunogen A 17 amino acid synthetic peptide near the carboxy terminus of human TTYH1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57348
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.