Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TTYH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562229
Description
Immunogen sequence: SDFCSNPDPY VLNLTQEETG LSSDILSYYL LCNRAVSNPF QQRLTLSQRA LANIHSQLLG LEREAVPQFP SAQKPLLSLE ETLNVTEGNF HQLVALLHCR SLHKDYGAAL Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
TTYH1 is a member of the tweety family of proteins, a family of chloride anion channels containing five transmembrane regions. TTYH1 is a Ca2+-independent, volume-sensitive large conductance chloride (Cl-) channel. TTYH1 is primarily expressed in neural tissue and upregulated in astrocytoma, glioma, and several other cancers. Recent experiments have shown that TTYH1 is an integral endoplasmic reticulum (ER) membrane protein involved in cell proliferation and is thought to play an essential role in embryonic cell growth, possibly through the Ca2+ storage/release process in ER membranes during early development.
Specifications
TTYH1 | |
Polyclonal | |
Unconjugated | |
TTYH1 | |
4930459B04Rik; 6330408P11Rik; hTTY1; mTTY1; Protein tweety homolog 1; tty; TTY1; TTYH1; tweety family member 1; tweety homolog 1; tweety homolog 1 (Drosophila) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57348 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H313 | |
TTYH1 | |
Recombinant protein corresponding to Human TTYH1. Recombinant protein control fragment (Product #RP-103083). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction