Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TTYH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562800
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562800 Supplier Invitrogen™ Supplier No. PA562800
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PWWVSLLHRL PHFDLSWEAT SSQFRPEDTD YQQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Probable large-conductance Ca(2+)-activated chloride channel. May play a role in Ca(2+) signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTYH3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TTYH3
Gene Accession No. Q9C0H2
Gene Alias 2900029G13Rik; AI414930; hTTY3; KIAA1691; mTTY3; Protein tweety homolog 3; TTY3; TTYH3; tweety 3; tweety family member 3; tweety homolog 3; tweety homolog 3 (Drosophila)
Gene Symbols TTYH3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TTYH3. Recombinant protein control fragment (Product #RP-98458).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80727
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.