Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUB 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981454
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1454 Supplier Bioss Supplier No. BS11536R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the Tubby family of bipartite transcription factors. The encoded protein may play a role in obesity and sensorineural degradation. The crystal structure has been determined for a similar protein in mouse, and it functions as a membrane-bound transcription regulator that translocates to the nucleus in response to phosphoinositide hydrolysis. Two transcript variants encoding distinct isoforms have been identified for this gene.

Specifications

Antigen TUB 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TUB
Gene Accession No. P50607
Gene Alias CELE_F10B5.4; F10B5.4; Rd5; RDOB; retinal degeneration 5; Tub; tub-1; tubby bipartite transcription factor; tubby candidate gene; tubby homolog; tubby homolog (mouse); tubby homologue; Tubby protein; Tubby protein homolog; Tubby protein homolog 1
Gene Symbols TUB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TUB 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7275
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.