Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUB Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-248 Supplier Invitrogen™ Supplier No. PA116934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TUB Polyclonal Antibody for Western Blot, IHC (P), IP

This gene encodes a member of the Tubby family of bipartite transcription factors. The encoded protein may play a role in obesity and sensorineural degradation. The crystal structure has been determined for a similar protein in mouse, and it functions as a membrane-bound transcription regulator that translocates to the nucleus in response to phosphoinositide hydrolysis. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUB
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene tub-1
Gene Accession No. Q09306
Gene Alias CELE_F10B5.4; F10B5.4; Rd5; RDOB; retinal degeneration 5; TUB; TUB bipartite transcription factor; TUB-1; tubby bipartite transcription factor; tubby candidate gene; tubby homolog; tubby homologue; tubby protein; Tubby protein homolog; Tubby protein homolog 1
Gene Symbols tub-1
Host Species Rabbit
Immunogen Recombinant C. elegans Tubby (domain residues 160-421).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 174312
Target Species C. elegans
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.