Learn More
Tuba Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303074A

Description
The recommended shelf life for this product is 1 year from date of receipt.
DNMBP belongs to the DBL (MIM 311030) family of guanine nucleotide exchange factors and plays a role in the regulation of cell junctions (Otani et al., 2006 [PubMed 17015620]).Specifications
Tuba | |
Polyclonal | |
Unconjugated | |
DNMBP | |
ARHGEF36, TUBA, dynamin-binding protein, scaffold protein TUBA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23268 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6XZF7 | |
DNMBP | |
Between 575 and 625 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.