Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBA1A/TUBA1B Polyclonal Antibody, Cyanine5, Bioss
SDP

Catalog No. 502280078
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0078 Supplier Bioss Supplier No. BS20496RCY5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TUBA1A/TUBA1B Polyclonal antibody specifically detects TUBA1A/TUBA1B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry

Specifications

Antigen TUBA1A/TUBA1B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine5
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene TUBA1B
Gene Accession No. P05213, P68363, P68369, P68370, Q6P9V9, Q71U36
Gene Alias TUBA1A; TUBA1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TUBA1A & TUBA1B (amino acids 401-451).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10376, 22142, 22143, 500929, 64158, 7846
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.