Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBA3E Monoclonal Antibody (OTI2A8), TrueMAB™, OriGene
SDP

Catalog No. 502542199
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2199 30 μL
50-167-2683 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2199 Supplier OriGene Supplier No. TA501545S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TUBA3E Monoclonal antibody specifically detects TUBA3E in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen TUBA3E
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene IVD
Gene Accession No. P12007, P26440
Gene Alias alpha-tubulin 3E; Detyrosinated tubulin alpha-3E chain; TUBA3E; tubulin alpha 3e; tubulin alpha-3E chain; tubulin, alpha 3e
Host Species Mouse
Immunogen Full length human recombinant protein of human IVD produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100856316, 24513, 3712
Target Species Canine, Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.