Learn More
Invitrogen™ TUBA4A Monoclonal Antibody (KT204)

Description
This product was used in an Indirect ELISA.
Specifications
Specifications
| Antigen | TUBA4A |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Monoclonal |
| Clone | KT204 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | TUBA4A |
| Gene Accession No. | P68366 |
| Gene Alias | Alpha-tubulin 1; Alpha-tubulin 4; alpha-tubulin isotype M-alpha-4; ALS22; H2-ALPHA; M[a]4; Testis-specific alpha-tubulin; TUBA1; Tuba4; TUBA4A; Tubulin alpha; tubulin alpha 4a; tubulin alpha-1 chain; Tubulin alpha-4 chain; tubulin alpha-4A chain; Tubulin H2-alpha; tubulin, alpha 1 (testis specific); tubulin, alpha 4; tubulin, alpha 4a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.