Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUBA6 Antibody (OTI1D8), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274692IR
Description
TUBA6 Monoclonal specifically detects TUBA6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TUBA6 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TUBA1C | |
Full length recombinant protein of human TUBA6 (NP_116093) produced in HEK293T cells. | |
0.1 mL | |
84790.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1D8 | |
Western Blot | |
Alpha-tubulin 6, bcm948, MGC10851, MGC14580, TUBA6, tubulin alpha-1C chain, Tubulin alpha-6 chain, tubulin, alpha 1c, tubulin, alpha 6 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction