Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBA8 Monoclonal Antibody (OTI3E10), TrueMAB™, OriGene
SDP

Catalog No. 502542609
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2609 30 μL
50-167-2152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2609 Supplier OriGene Supplier No. TA501009S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TUBA8 Monoclonal antibody specifically detects TUBA8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TUBA8
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3E10
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TLDC1
Gene Accession No. Q6P9B6
Gene Alias alpha tubulin 8; alpha-tubulin 8; TUBA8; TUBAL2; tubulin alpha 8; tubulin alpha chain-like 2; Tubulin alpha-8 chain; tubulin, alpha 8
Host Species Mouse
Immunogen Full length human recombinant protein of human KIAA1609 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57707
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.