Learn More
TUBA8 Mouse anti-Human, Clone: OTI3E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501009

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the alpha tubulin protein family. Alpha tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Mutations in this gene are associated with polymicrogyria and optic nerve hypoplasia. Alternate splicing results in multiple transcript variants.Specifications
TUBA8 | |
Monoclonal | |
Unconjugated | |
TUBA8 | |
CDCBM8, TUBAL2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
51807 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NY65 | |
TUBA8 | |
Full length human recombit protein of human TUBA8 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.